Learn More
Sodium acetate, anhydrous, ACS, 99.0% min
Description
Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 324°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Assay Percent Range | 99.0% min. |
| Beilstein | 3595639 |
| Solubility Information | Soluble in water,ethanol,methanol and hydrazine. |
| Formula Weight | 82.03 |
| Grade | ACS |
| Odor | Nearly odorless |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H500 |
| EINECSNumber | 204-823-8 |
| RTECSNumber | AJ4300010 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.