Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, Anhydrous, Purified, Reagents

Catalog No. 01337067
Change view
Click to view available options
Quantity:
500 g
3 kg
Catalog No. Quantity
01-337-067 3 kg
01-337-066 500 g
2 options

Catalog No. 01-337-067

Supplier: Reagents Holdings Llc C23064003KG

Only null left
Add to Cart
Add to Cart

Sodium Acetate, Anhydrous, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Boiling Point 123°C
Melting Point 324°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.528
Chemical Name or Material Sodium Acetate, Anhydrous, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.