Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Allylsulfonate 94.0+%, TCI America™
SDP

Catalog No. A080925G
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 2495-39-8
Molecular Formula C3H5NaO3S
Molecular Weight (g/mol) 144.12
MDL Number MFCD00051416
InChI Key DIKJULDDNQFCJG-UHFFFAOYSA-M
Synonym sodium allylsulfonate, sodium prop-2-ene-1-sulfonate, 2-propene-1-sulfonic acid, sodium salt, sodium allyl sulfonate, sodium 2-propene-1-sulfonate, unii-7sgf7tb9o2, sodium prop-2-enesulphonate, allylsulfonic acid sodium salt, sodium allysulfonate, 2-propene-1-sulfonic acid sodium salt
PubChem CID 23690996
IUPAC Name sodium;prop-2-ene-1-sulfonate
SMILES C=CCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 220°C
Color White
Quantity 25 g
Formula Weight 144.12
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Allylsulfonate
EINECSNumber (2)-1590
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.