Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cacodylate, 0.1M buffer soln., pH 6.5

Catalog No. aaj60367ad
Click to view available options
Quantity:
50 mL
100 mL

CAS: 6131-99-3214.024 g/mol

Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (∼100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-99-3
Molecular Weight (g/mol) 214.024

Specifications

Concentration 0.1M buffer soln.
pH 6.5
Physical Form Liquid
Quantity 50 mL
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.