Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Calcium Exchanger 1/NCX1 Antibody (SR1046), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32188650UL
Description
Sodium Calcium Exchanger 1/NCX1 Monoclonal antibody specifically detects Sodium Calcium Exchanger 1/NCX1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Sodium Calcium Exchanger 1/NCX1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1046 | |
Western Blot 1:500-1:2000 | |
FLJ37694, FLJ43417, Na(+)/Ca(2+)-exchange protein 1, sodium/calcium exchanger 1, solute carrier family 8 (sodium/calcium exchanger), member 1 | |
A synthesized peptide derived from human Sodium Calcium Exchanger 1/NCX1 (Uniprot #: P32418) | |
50 μg | |
Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
6546 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction