Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Calcium Exchanger 1/NCX1 Antibody (SR1046), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$309.50 - $423.00

Specifications

Antigen Sodium Calcium Exchanger 1/NCX1
Clone SR1046
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB258490
SDP
View Documents
Novus Biologicals
NBP321886100UL
100 μg
Each for $423.00
Only null left
Add to Cart
 
NB260510
SDP
View Documents
Novus Biologicals
NBP32188650UL
50 μg
Each for $309.50
Only null left
Add to Cart
 
Description

Description

Sodium Calcium Exchanger 1/NCX1 Monoclonal antibody specifically detects Sodium Calcium Exchanger 1/NCX1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Sodium Calcium Exchanger 1/NCX1
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
FLJ37694, FLJ43417, Na(+)/Ca(2+)-exchange protein 1, sodium/calcium exchanger 1, solute carrier family 8 (sodium/calcium exchanger), member 1
A synthesized peptide derived from human Sodium Calcium Exchanger 1/NCX1 (Uniprot #: P32418)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1046
Western Blot
Unconjugated
Rabbit
Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
6546
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.