Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium-Calcium Exchanger Antibody (C2C12), Novus Biologicals™
SDP

Catalog No. NB1202869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202869 Supplier Novus Biologicals Supplier No. NB1202869
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Sodium-Calcium Exchanger Monoclonal specifically detects Sodium-Calcium Exchanger in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Insect, Kangaroo, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Sodium-Calcium Exchanger
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone C2C12
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100
Gene Alias Na(+)/Ca(2+)-exchange protein 3, Na+/Ca2+ exchanger 3, NCX3sodium/calcium exchanger SLC8A3, sodium/calcium exchanger 3, solute carrier family 8 (sodium/calcium exchanger), member 3, solute carrier family 8 (sodium-calcium exchanger), member 3
Gene Symbols SLC8A3
Host Species Mouse
Immunogen Purified canine cardiac sodium/calcium exchanger.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6547
Test Specificity Sodium Calcium Exchanger (C2C12)
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Insect, Kangaroo, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.