Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate decahydrate, 99+%, for analysis

Catalog No. p-41842
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 6132-02-1 | CNa2O3·10H2O | 286.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-02-1
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34°C
Color White
Physical Form Small Crystals
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Fieser 07,322
Merck Index 15, 8731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol
Formula Weight 286.14
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.46
Density 1.4600g/mL
Chemical Name or Material Sodium carbonate decahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.