Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate Tetrahydrate, Reagents

Catalog No. 01337031
Change view
Click to view available options
:
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-031 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-031 Supplier Reagents Holdings Llc Supplier No. C2314050100C2
Only null left
Add to Cart
Add to Cart

Sodium Chromate Tetrahydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-82-9
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 161.97
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym Chromium Disodium Oxide Tetrahydrate
IUPAC Name disodium dioxochromiumbis(olate)
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Color Yellow
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.73
Chemical Name or Material Sodium Chromate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.