Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanate, 95%

Catalog No. p-7082118
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 917-61-3 | CNNaO | 65.007 g/mol

Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

Solubility
Soluble in water 11.6 g/100 mL (25°C). Slightly soluble in ammonia, benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-61-3
Molecular Formula CNNaO
Molecular Weight (g/mol) 65.007
MDL Number MFCD00011121
InChI Key ZVCDLGYNFYZZOK-UHFFFAOYSA-M
Synonym sodium cyanate, zassol, cyanic acid, sodium salt, sodiumcyanate, cyansan, weecon, sodium isocyanate, san-cyan, unii-8ufs3jrv8p, naocn
PubChem CID 517096
ChEBI CHEBI:38906
IUPAC Name sodium;cyanate
SMILES C(#N)[O-].[Na+]

Specifications

Melting Point 550°C
Density 1.89
Quantity 100 g
Beilstein 3594963
Merck Index 14,8604
Solubility Information Soluble in water 11.6g/100 mL (25°C). Slightly soluble in ammonia,benzene. Insoluble in diethyl ether.
Formula Weight 65.01
Percent Purity 95%
Chemical Name or Material Sodium cyanate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-030-6
RTECSNumber GS7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.