Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Cyanide (Granular/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Specifications
Specifications
| Boiling Point | 1497°C |
| Melting Point | 562°C |
| Color | White |
| pH | 11 to 12 |
| Physical Form | Solid |
| Quantity | 100 g |
| Formula Weight | 49.01 |
| Percent Purity | ≥95% |
| Grade | Certified ACS |
| Identification | Pass Test |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 6.1, : Poison |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction