Learn More
Sodium D-gluconate, 97%
Description
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.
Solubility
Very soluble in water; sparingly soluble in alcohol; insoluble in ether.
Notes
In general it is stable, non corrosive, non-toxic, readily biodegradable and incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽206°C (decomposition) |
| Quantity | 2500 g |
| Beilstein | 3919651 |
| Solubility Information | Very soluble in water; sparingly soluble in alcohol; insoluble in ether. |
| Formula Weight | 218.14 |
| Chemical Name or Material | Sodium D-gluconate |
Safety and Handling
| EINECSNumber | 208-407-7 |
| RTECSNumber | LZ5235000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.