Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Erythorbate, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606499 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

S1223, 6381-77-7, C6H7NaO6·H2O

Spectrum™ Chemical Sodium Erythorbate, FCC is used as an antioxidant preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6381-77-7
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key QNSCGSPRKIPKTN-RKJRWTFHSA-N
IUPAC Name sodium (2R)-2-[(2R)-4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl]-2-hydroxyethan-1-olate
SMILES [Na+].O[C@H](C[O-])[C@H]1OC(O)=C(O)C1=O

Specifications

CAS Max % 1
Loss on Drying 0.0025
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.