Learn More
Sodium fluoride, 99%, Thermo Scientific Chemicals
Description
Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.
Solubility
Slightly soluble in water. Insoluble in alcohol
Notes
It must be kept away from oxidizing agents, metals, acids, alkalis. It is less hygroscopic.
Specifications
Specifications
| Boiling Point | 1704°C |
| Melting Point | 993°C |
| Quantity | 1000 g |
| Linear Formula | NaF |
| UN Number | UN1690 |
| Merck Index | 14,8618 |
| Solubility Information | Slightly soluble in water. Insoluble in alcohol |
| IUPAC Name | sodium;fluoride |
| Formula Weight | 41.99 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H301-H315-H319 |
| Hazard Statement | GHS H Statement H301-H315-H319 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM FLUORIDE, SOLID |
| EINECSNumber | 231-667-8 |
| RTECSNumber | WB0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.