Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium formaldehydesulfoxylate dihydrate, 98%

Catalog No. AC149190010
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle

Catalog No. AC149190010

Supplier: Thermo Scientific Chemicals 149190010

Only null left
Add to Cart

CAS: 6035-47-8 | CH3NaO32H2O | 154.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6035-47-8
Molecular Formula CH3NaO32H2O
Molecular Weight (g/mol) 154.11
MDL Number MFCD00150598
InChI Key UCWBKJOCRGQBNW-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate
PubChem CID 23666330
IUPAC Name sodium;hydroxymethanesulfinate;dihydrate
SMILES C(O)S(=O)[O-].O.O.[Na+]

Specifications

Melting Point 63.0°C to 64.0°C
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula HOCH2SO2Na·2H2O
Quantity 1 kg
Merck Index 15,8755
Solubility Information Solubility in water: 1088g/L. Other solubilities: practically insoluble in abs.alcohol, ether and, benzene
Formula Weight 154.11
Percent Purity 98%
Chemical Name or Material Sodium formaldehydesulfoxylate dihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
Suspected of damaging the unborn child.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.