Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluoroantimonate(V), 99.5%

Catalog No. p-4407889
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1306614 25 g
AA1306622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1306614 Supplier Thermo Scientific Chemicals Supplier No. 01306614
Only null left
Add to Cart

CAS: 16925-25-0 | F6NaSb | 258.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16925-25-0
Molecular Formula F6NaSb
Molecular Weight (g/mol) 258.74
MDL Number MFCD00003482
InChI Key HKLMYZVMEYYVBS-UHFFFAOYSA-H
Synonym sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt
PubChem CID 16689647
IUPAC Name sodium;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Na+]

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.5%
Formula Weight 258.73
Odor Odorless
Appearance White powder
Density 3.38
Chemical Name or Material Sodium hexafluoroantimonate(V)
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1, Packing Group: III
EINECSNumber 240-989-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.