Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluorosilicate, >99%, Thermo Scientific Chemicals

Catalog No. p-4407890
Click to view available options
Quantity:
500 g
2 kg

Sodium hexafluorosilicate is used as a additive for water fluoridation, opal glass raw material and in ore refining. It is also used in the preparation of other fluoride chemicals like sodium fluoride, sodium hexafluoroaluminate, potassium hexafluorosilicate, ammonium fluoride and calcium fluoride. It finds application as flotator, component for wet etching, wood preservative, brazing flux and welding compound. It is an active component of concrete.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16893-85-9
Molecular Formula F6Na2Si
Molecular Weight (g/mol) 188.06
MDL Number MFCD00003491
InChI Key TWGUZEUZLCYTCG-UHFFFAOYSA-N
Synonym sodium hexafluorosilicate, sodium fluosilicate, salufer, sodium silicofluoride, safsan, super prodan, destruxol applex, ortho earwig bait, ortho weevil bait, ens-zem weevil bait
PubChem CID 28127
SMILES [Na+].[Na+].F[Si--](F)(F)(F)(F)F

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range >99%
Solubility Information Slightly soluble in water. Solubility increases with temperature. Insoluble in alcoholSlightly soluble in water. Insoluble in alcohol.
IUPAC Name disodium hexafluorosilanediuide
Formula Weight 188.06
Refractive Index 1.31
Density 2.679 g/cm3 at 20°C
Chemical Name or Material Sodium hexafluorosilicate
Show More Show Less
Hazard Category H301+H311+H331
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 240-934-8
RTECSNumber VV8410000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.