Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.04 N in Methanol, Reagents

Catalog No. 01333955
Change view
Click to view available options
:
Quantity: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-955 Quantity: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-955 Supplier Reagents Holdings Llc Supplier No. CS012700100A
Only null left
Add to Cart
Add to Cart

Sodium Hydroxide, 0.04 N in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 1310-73-2 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Boiling Point 65°C
Quantity 100 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8029
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.04 N in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.