Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Iodide Symporter Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 243241AP150UL
Description
This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extra membranous domains. It may represent an autoantigen in thyroid.Specifications
Sodium Iodide Symporter | |
Polyclonal | |
Unconjugated | |
SLC5A5 | |
NIS, SLC5A5, Sodium iodide symporter, Sodium/iodide cotransporter | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114479, 114613, 6528 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q63008, Q92911, Q99PN0 | |
SLC5A5 | |
Sodium iodide symporter Fusion Protein Ag19504 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction