Alfa Aesar™ Sodium iodide, ultra dry, 99.98% (metals basis)

Manufacturer: Alfa Aesar™ 4344714

Quantity 25g

 View more versions of this product

Catalog No. AA4344714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7681-82-5
    Formula Weight 149.89
    MDL Number MFCD00003532
    Merck Index 14,8631
    Chemical Name or Material Sodium iodide, Ultra dry

    Specifications

    Formula Weight 149.89
    Boiling Point 1300° to 1304°C
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −10 Mesh
    Refractive Index 1.7745
    Sensitivity Hygroscopic
    MDL Number MFCD00003532
    Melting Point 651°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    CAS 7681-82-5
    Merck Index 14,8631
    Health Hazard 1 Warning
    Density 3.67g/mL
    TSCA TSCA
    Physical Form Beads
    Quantity 25g
    Storage Note 1
    • Ampouled under argon
    RTECS Number WB6475000
    Chemical Name or Material Sodium iodide, Ultra dry
    Assay 99.98%, (metals basis)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit