Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium L-Lactate, Approx. 98%, Spectrum™ Chemical
SDP

Catalog No. 18606631 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle

S2008, 867-56-1, C3H5NaO3

Spectrum™ Chemical Sodium L-Lactate , also known as milk acid, is used in various biochemical processes. It is a carboxylic acid and has a hydroxyl group adjacent to the carboxyl group. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
InChI Key NGSFWBMYFKHRBD-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxypropanoate
SMILES [Na+].CC(O)C([O-])=O

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity ∼98%
Grade Reagent
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.