Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 255.0°C to 273.0°C |
| Quantity | 1 kg |
| Assay Percent Range | 97.5% min. (Complexometry) |
| Merck Index | 15,1619 |
| Formula Weight | 230.5 |
| Percent Purity | 98% |
| Grade | Pure |
| Loss on Drying | 0.5% max. |
| Chemical Name or Material | Cadmium acetate |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause cancer. |
| Precautionary Statement | GHS P Statement Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortab |
| EINECSNumber | 208-853-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction