Learn More
Sodium nitrite, ACS, 97% min
Description
Sodium nitrite is used in many industrial applications like meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, as an antidote in cyanide poisoning, as an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 271°C |
| Physical Form | Granules |
| Quantity | 500 g |
| Assay Percent Range | 97% min |
| Solubility Information | Soluble in water,methanol,ethanol,ammonia and nitric acid. Slightly soluble diethyl ether. |
| Formula Weight | 69 |
| Grade | ACS Reagent |
| Sensitivity | Hygroscopic |
| Density | 2.17 g/mL |
| Chemical Name or Material | Sodium nitrite, For ACS analysis; Decomposes above 320° |
Safety and Handling
| Hazard Category | H272-H301-H319-H373 |
| Precautionary Statement | P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: III |
| EINECSNumber | 231-555-9 |
| RTECSNumber | RA1225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.