Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium peroxide, 95%

Catalog No. p-7046149
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 1313-60-6 | Na2O2 | 77.978 g/mol

Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

Solubility
Soluble in acid. Insoluble in alkali.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. It reacts with water violently to give sodium hydroxide and hydrogen peroxide. Incompatible with strong reducing agents, organic materials, alcohols and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 460°C (decomposition)
Quantity 50 g
UN Number UN1504
Merck Index 14,8655
Solubility Information Soluble in acid. Insoluble in alkali.
Formula Weight 77.98
Percent Purity 95%
Sensitivity Air sensitive; Hygroscopic
Density 2.805
Chemical Name or Material Sodium peroxide
Hazard Category H271-H290-H314-H335
Hazard Statement GHS H Statement
H271-H314
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: SODIUM PEROXIDE
EINECSNumber 215-209-4
RTECSNumber WD3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.