Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Peroxide, Reagent, ACS, 93%, Spectrum™ Chemical
SDP

Catalog No. 18606695 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
Steel Can

Catalog No. 18-606-695

Supplier: Spectrum Chemical Manufacturing Corporation S1390500GM

Only null left
Add to Cart
Add to Cart

S1390, 1313-60-6, Na2O2

Spectrum™ Chemical Sodium Peroxide, Reagent, ACS is a solid that comes in the form yellow micro beads. It is an inorganic compound that was historically used to bleach wood pulp in order to prepare it for the use in paper manufacture. Today it is most commonly used for certain specialized lab operations such as mineral extraction form ore. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.98
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
IUPAC Name disodium dioxidanediide
SMILES [Na+].[Na+].[O-][O-]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 93%
Grade ACS
Packaging Steel Can
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.