Learn More
Rhenium(VI) oxide, 99.9% (metals basis)
Description
Rhenium(VI) oxide is involved as a catalyst for the hydrogenation of carboxylic acids and carboxamides. It is involved in the alkene epoxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 400°C (decomposition) |
| Physical Form | Powder |
| Quantity | 1 g |
| Assay Percent Range | 99.9% (metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 236.2 |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Rhenium(VI) oxide |
Safety and Handling
| EINECSNumber | 215-228-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.