Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, 0.2 M Buffer Solution, pH 8.0, Low Endotoxin, PROMO, Thermo Scientific Chemicals

Catalog No. J67215AKPM
Click to view available options
Quantity:
250 mL

Chemical Identifiers

CAS 7732-18-5

Specifications

Color Colorless
Concentration 0.2 M
pH 8.0
Quantity 250 mL
Form Liquid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.