Learn More
Sodium phosphonoformate hexahydrate, 98+%
Description
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| IUPAC Name | trisodium;phosphonatoformate;hexahydrate |
| Formula Weight | 300.08 (191.98 Anhydrous) |
| Percent Purity | ≥98% |
| Chemical Name or Material | Sodium phosphonoformate hexahydrate |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation |
| RTECSNumber | SY8200000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.