Learn More
Sodium Potassium ATPase Alpha 1 Antibody (464.6), Janelia Fluor™ 646, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Sodium Potassium ATPase Alpha 1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 464.6 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | ATPase, Na+/K+ transporting, alpha 1 polypeptide, EC 3.6.3, EC 3.6.3.9, K-ATPase alpha-1 subunit, K-ATPase catalytic subunit alpha-A protein, MGC3285, MGC51750, Na, Na(+)/K(+) ATPase alpha-1 subunit, Na, K-ATPase, alpha-A catalytic polypeptide, Na+, K+ ATPase alpha subunit, Na+/K+ ATPase 1, sodium pump 1, Sodium pump subunit alpha-1, sodium/potassium-transporting ATPase subunit alpha-1, sodium-potassium-ATPase, alpha 1 polypeptide |
| Gene Symbols | ATP1A1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.