Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium ATPase Alpha 3 Antibody (XVIF9-G10), PE, Novus Biologicals™
SDP

Catalog No. NB300540PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300540PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300540PE Supplier Novus Biologicals Supplier No. NB300540PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sodium Potassium ATPase Alpha 3 Monoclonal specifically detects Sodium Potassium ATPase Alpha 3 in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Flow Cytometry.

Specifications

Antigen Sodium Potassium ATPase Alpha 3
Applications Flow Cytometry
Classification Monoclonal
Clone XVIF9-G10
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. P13637
Gene Alias alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3
Gene Symbols ATP1A3
Host Species Mouse
Immunogen Canine cardiac microsomes.
Molecular Weight of Antigen 111.749 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 478
Test Specificity Detects sodium potassium ATPase. This is specific for the alpha-3 subunit.
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.