Learn More
Sodium Potassium ATPase Beta 1 Antibody (464.8 (also known as 8A)), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Sodium Potassium ATPase Beta 1 |
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 464.8 (also known as 8A) |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin reported in scientific literature |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | Q9TT37 |
| Gene Alias | adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1 |
| Gene Symbols | ATP1B1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.