Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Potassium ATPase Beta 1 Antibody (464.8 (also known as 8A)), Novus Biologicals™
SDP

Catalog No. NB300147SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300147SS 0.025 mL
NB300147 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300147SS Supplier Novus Biologicals Supplier No. NB300147SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 14 publications

Sodium Potassium ATPase Beta 1 Monoclonal specifically detects Sodium Potassium ATPase Beta 1 in Bovine, Canine, Chicken, Hamster, Human, Mouse, Mouse (Negative), Pig, Rabbit, Rat, Xenopus species, Xenopus species (African Clawed Frog) samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Sodium Potassium ATPase Beta 1
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 464.8 (also known as 8A)
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin reported in scientific literature
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q9TT37
Gene Alias adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1
Gene Symbols ATP1B1
Host Species Mouse
Immunogen Purified Sodium Potassium ATPase Beta 1 from rabbit renal outer medulla. [UniProt# Q9TT37]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 481
Test Specificity This is specific for Na,K-ATPase beta 1 subunit.
Target Species Bovine, Canine, Chicken, Hamster, Human, Mouse, Mouse (Negative), Pig, Rabbit, Rat, Xenopus species, Xenopus species
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.