Learn More
Sodium sulfate, anhydrous, 99.99% (metals basis)
Description
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 884°C |
| Color | White |
| Physical Form | Chunks and/or Powder |
| Quantity | 100 g |
| Linear Formula | Na2SO4 |
| Merck Index | 14,8680 |
| Solubility Information | Soluble in water,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents. |
| Formula Weight | 142.04 |
| Percent Purity | 99.99% |
| Odor | Odorless |
| Show More |
Safety and Handling
| EINECSNumber | 231-820-9 |
| RTECSNumber | WE1650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.