Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS0850100 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
White Poly Wide Mouth Bottle

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 1 kg
Percent Purity ≥ 99%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Sulfate, Anhydrous
ShelfLife 60 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.