Learn More
Sodium sulfide nonahydrate, ACS, 98.0% min, Thermo Scientific Chemicals
Description
It is the precursor material for preparation of cadmium sulfide quantum dots. Also employed in the pulp and paper industry. It is likewise employed in the making of colors and dyestuffs. It is too useful in the trace quantities for the rapid synthesis of small silver nanocubes through mediating polyol reduction. It has as a significant role in breaking up the feather keratin useful for the preparation of bio-polymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽50°C (decomposition) |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Assay Percent Range | 98% min. |
| Solubility Information | Soluble in water; slightly soluble in alcohol. Insoluble in ether. |
| Formula Weight | 240.18 (78.04 Anhydrous) |
| Grade | ACS |
| Sensitivity | Hygroscopic; Light sensitive |
| Appearance | Crystalline |
| Density | 1.427 g/cm3 at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H290-H302-H311-H314 |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 215-211-5 |
| RTECSNumber | WE1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.