Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium trifluoromethanesulfonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2926-30-9
CF3NaO3S
172.05
MFCD00061607
XGPOMXSYOKFBHS-UHFFFAOYSA-M
sodium trifluoromethanesulfonate,sodium triflate,trifluoromethanesulfonic acid sodium salt,sodium trifluoro methane sulfonate,methanesulfonic acid, trifluoro-, sodium salt,pubchem23598,cf3so3na,acmc-1cfau,sodiumtrifluoromethanesulfonate,ksc491a1d
23676748
sodium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[Na+]
2926-30-9
CF3NaO3S
172.05
MFCD00061607
XGPOMXSYOKFBHS-UHFFFAOYSA-M
sodium trifluoromethanesulfonate,sodium triflate,trifluoromethanesulfonic acid sodium salt,sodium trifluoro methane sulfonate,methanesulfonic acid, trifluoro-, sodium salt,pubchem23598,cf3so3na,acmc-1cfau,sodiumtrifluoromethanesulfonate,ksc491a1d
23676748
sodium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[Na+]
Packaging Glass bottle
Melting Point 253.0°C to 255.0°C
Quantity 5g
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula F3COSNa
Solubility Information Solubility in water: soluble in water.
Formula Weight 172.05
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Sodium trifluoromethanesulfonate, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.