Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SoftGUARD™ Extra-Strength Barrier Hand Cream

Catalog No. p-4525623 Shop All Epredia Products
Click to view available options
Volume (English):
3 oz.
8 oz.
16 oz.
1 gal.
Packaging:
Pump Bottle
Refill Jug
Tube
This item is not returnable. View return policy
This item is not returnable. View return policy

Prevent skin irritation caused by frequent washing and gloving with Epredia™ SoftGUARD Extra-Strength Barrier Hand Cream.

Unique barrier feature helps prevent the occlusion of the skin.
  • Compatible with latex, nitrile, vinyl, PCMX, CHG, and antimicrobial soap—petroleum- and mineral oil-free
  • Utilizes an extra-strength antimicrobial preservative to prevent cross-contamination
  • Absorbs immediately
  • Nongreasy, fragrance-free and pH balanced with the skin

Specifications

Product Type Extra-strength Barrier Hand Cream
Packaging Pump Bottle
Volume (English) 16 oz.
Volume (Metric) 473 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.