Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solanezumab Antibody (2372A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340187 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340187 Supplier Novus Biologicals Supplier No. FAB10124MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Solanezumab Monoclonal antibody specifically detects Solanezumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Solanezumab
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2372A
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Full-length Solanezumab
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.