Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Solid Penny-Head Stopper



Flask length solid penny head stoppers
$22.56 - $41.22
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Organic Synthesis |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity & Availability | |||||
31-501-876
![]() ![]() |
Synthware™
S291018 |
10/18 |
Each for $22.56
|
|
|||||
31-501-877
![]() ![]() |
Synthware™
S291030 |
10/30 |
Each for $22.87
|
|
|||||
31-501-878
![]() ![]() |
Synthware™
S291420 |
14/20 |
Each for $24.12
|
|
|||||
31-501-879
![]() ![]() |
Synthware™
S291922 |
19/22 |
Each for $28.02
|
|
|||||
31-501-880
![]() ![]() |
Synthware™
S292440 |
24/40 |
Each for $34.80
|
![]() |
|||||
31-501-881
![]() ![]() |
Synthware™
S292942 |
29/42 |
Each for $41.22
|
|
|||||
Specifications
Clear | |
Borosilicate Glass | |
Organic Synthesis | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title