Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soluble Liver/Pancreas Antigen Antibody, Novus Biologicals™
SDP

Catalog No. NBP220454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220454 Supplier Novus Biologicals Supplier No. NBP220454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Soluble Liver/Pancreas Antigen Polyclonal specifically detects Soluble Liver/Pancreas Antigen in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Soluble Liver/Pancreas Antigen
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9HD40
Gene Alias EC 2.9.1.2, EC 2.9.1.n1, Liver-pancreas antigen, LPDKFZp434B1417, MGC161491, O-phosphoseryl-tRNA(Sec) selenium transferase, Sec synthase, Selenocysteine synthase, Selenocysteinyl-tRNA(Sec) synthase, Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase, SepSecS, Sep-tRNA:Sec-tRNA synthase, SLA/LP, SLA/LP autoantigen, SLA-p35, SLAUGA suppressor tRNA-associated protein, Soluble liver antigen, soluble liver antigen/liver pancreas antigen, tRNA(Ser/Sec)-associated antigenic protein, TRNP48
Gene Symbols SEPSECS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLA/LP. The exact sequence is proprietary.
Molecular Weight of Antigen 56 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51091
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.