Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solute carrier family 22 member 18 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Solute carrier family 22 member 18 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:20 - 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Solute carrier family 22 member 18 Polyclonal specifically detects Solute carrier family 22 member 18 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Solute carrier family 22 member 18 | |
Polyclonal | |
Rabbit | |
Human | |
BWR1AORCTL2, BWSCR1Asolute carrier family 22 (organic cation transporter), member 1-like, Efflux transporter-like protein, HET, imprinted multi-membrane spanning polyspecific transporter-related protein 1, Imprinted multi-membrane-spanning polyspecific transporter-related protein 1, IMPT1DKFZp667A184, ITMp45-BWR1A, ORCTL-2, organic cation transporter-like 2, Organic cation transporter-like protein 2, p45 Beckwith-Wiedemann region 1A, SLC22A1Lcandidate A, solute carrier family 22 member 18, Solute carrier family 22 member 1-like, solute carrier family 22, member 18, TSSC5p45-Beckwith-Wiedemann region 1 A, Tumor-suppressing STF cDNA 5 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein | |
SLC22A18 | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5002 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PASTKGAKTDAQAPLPGGPRASVFDLKAIASLLRLPDVPRI | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title