Learn More
Honeywell™ Solutions for Eyesaline Porta-Stream Portable Eyewash Stations
Shop All Protective Industrial Products Inc. ProductsDescription
Available in concentrate/Saline form to refill bottles in Porta Stream™ wall-mounted stations or to replace untreated water in other portable eyewash equipment.
- Concentrate 70 oz. (2L) bottle makes about 6 gal. (22.8L)
- Concentrate 180 oz. (5.3L) bottle makes about 16 gal. (60.6L)
- Saline 128 oz. (3.8L) is ready-to-use solution
Specifications
Specifications
| Product Type | Saline Concentrate |
| Description | Saline Concentrate |
| Volume (English) | 180 oz. |
| Volume (Metric) | 5323.23 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.