Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin Antibody (YC7), Novus Biologicals™

Rat Monoclonal Antibody has been used in 2 publications
$192.47 - $519.67
Specifications
| Antigen | Somatostatin |
|---|---|
| Clone | YC7 |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017624
![]() |
Novus Biologicals
NB100646500.025ML |
0.025 mL |
Each for $192.47
|
|
|||||
NB10064650
![]() |
Novus Biologicals
NB10064650 |
0.1 mL |
Each for $519.67
|
|
|||||
Description
Somatostatin Monoclonal specifically detects Somatostatin in Human, Mouse, Rat, Porcine, Guinea Pig, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Somatostatin | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Pig, Rat, Zebrafish, Mouse, Guinea Pig, Human | |
| P61278 | |
| 6750 | |
| Somatostatin conjugated to thyroglobulin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| YC7 | |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| Tissue culture supernatant with 0.05% Thimerosal | |
| Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
| SST | |
| IgG2b | |
| Tissue culture supernatant | |
| NB100-64650 is specific for somatostatin. A hormone involved in endocrine system regulation through interactions with pituitary growth hormone, thyroid stimulating hormone, and a number of hormones of the gastrointestinal tract. Somatostatin has active 14 amino acid and 28 amino acid forms, produced by alternate cleavage of the gene product.This antibody does not cross react with enkephalins, other endorphins, substance P, or CGRP. It partially cross reacts with somatostatin fragments. The epitope recognized by this antibody lies within the 1-14 amino acid sequence of cyclic somatostatin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title