Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin Antibody (YC7), Novus Biologicals™
SDP

Catalog No. NB10064650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064650 0.1 mL
NB017624 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064650 Supplier Novus Biologicals Supplier No. NB10064650
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 2 publications

Somatostatin Monoclonal specifically detects Somatostatin in Human, Mouse, Rat, Porcine, Guinea Pig, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Somatostatin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YC7
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Tissue culture supernatant with 0.05% Thimerosal
Gene Accession No. P61278
Gene Alias Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28
Gene Symbols SST
Host Species Rat
Immunogen Somatostatin conjugated to thyroglobulin.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6750
Test Specificity NB100-64650 is specific for somatostatin. A hormone involved in endocrine system regulation through interactions with pituitary growth hormone, thyroid stimulating hormone, and a number of hormones of the gastrointestinal tract. Somatostatin has active 14 amino acid and 28 amino acid forms, produced by alternate cleavage of the gene product.This antibody does not cross react with enkephalins, other endorphins, substance P, or CGRP. It partially cross reacts with somatostatin fragments. The epitope recognized by this antibody lies within the 1-14 amino acid sequence of cyclic somatostatin.
Target Species Human, Mouse, Rat, Pig, Guinea Pig, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.