Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R1/SSTR1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Somatostatin R1/SSTR1
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227413SS
SDP
View Documents
Novus Biologicals
NBP227413SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227413
SDP
View Documents
Novus Biologicals
NBP227413
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Somatostatin R1/SSTR1 Polyclonal specifically detects Somatostatin R1/SSTR1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Somatostatin R1/SSTR1
Western Blot 1:100-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1
SSTR1
IgG
Protein G purified
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
6751
This antibody was raised against Somatostatin Receptor 1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.