Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R1/SSTR1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300120 0.1 mL
NB300120SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300120 Supplier Novus Biologicals Supplier No. NB300120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Somatostatin R1/SSTR1 Polyclonal specifically detects Somatostatin R1/SSTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Somatostatin R1/SSTR1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation 1:10-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P28646
Gene Alias G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1
Gene Symbols SSTR1
Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of rat Somatostatin Receptor 1, conjugated to cysteine. [UniProt# P28646]
Molecular Weight of Antigen 60 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6751
Test Specificity Specific to Somatostatin Receptor I (SSR1) and does not react with Somatostatin Receptor II (SSR2).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.