Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R2/SSTR2 Antibody (402038), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB370651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370651 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370651 Supplier Novus Biologicals Supplier No. FAB4224Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Somatostatin R2/SSTR2 Monoclonal specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen Somatostatin R2/SSTR2
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 402038
Conjugate DyLight 680
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with Somatostatin R2/SSTR2, Met1-Ile369, Accession # P30874
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6752
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.