Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R2/SSTR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300157SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300157SS 0.025 mL
NB300157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300157SS Supplier Novus Biologicals Supplier No. NB300157SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

Somatostatin R2/SSTR2 Polyclonal specifically detects Somatostatin R2/SSTR2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.

Specifications

Antigen Somatostatin R2/SSTR2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:1000, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Electron Microscopy
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P30874, P30875, P30680
Gene Alias somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R
Gene Symbols SSTR2
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of rat Somatostatin Receptor 2 [UniProt# P30680]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6752
Target Species Human, Mouse, Rat, Pig
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.