Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R2/SSTR2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 9 publications

$243.50 - $489.50

Specifications

Antigen Somatostatin R2/SSTR2
Dilution Western Blot 1:100 - 1:1000, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Electron Microscopy
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300157SS
SDP
View Documents
Novus Biologicals
NB300157SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB300157
SDP
View Documents
Novus Biologicals
NB300157
0.1 mL
Each for $489.50
Only null left
Add to Cart
 
Description

Description

Somatostatin R2/SSTR2 Polyclonal specifically detects Somatostatin R2/SSTR2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.
Specifications

Specifications

Somatostatin R2/SSTR2
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Unconjugated
Rabbit
Alzheimers Research, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Whole antisera with 0.05% Sodium Azide
somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R
SSTR2
IgG
Unpurified
Western Blot 1:100 - 1:1000, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Electron Microscopy
Polyclonal
Antisera
RUO
Human, Mouse, Rat, Pig
P30874, P30875, P30680
6752
A synthetic peptide made to the C-terminus of rat Somatostatin Receptor 2 [UniProt# P30680]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.