Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R3/SSTR3 Antibody (576017), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB375296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375296 Supplier Novus Biologicals Supplier No. FAB7018L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Somatostatin R3/SSTR3 Monoclonal specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Somatostatin R3/SSTR3
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 576017
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias somatostatin receptor 3, SS-3-R, SS3-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Somatostatin R3/SSTR3, Accession # P32745
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6753
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.